missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 4-(trifluoromethyl)phenylacetate, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H64099MD
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ethyl 4-(trifluoromethyl)phenylacetate | |
Colorless | |
MFCD07779351 | |
ethyl 2-4-trifluoromethyl phenyl acetate, ethyl 4-trifluoromethyl phenylacetate, 4-trifluoromethyl benzeneacetic acid ethyl ester, 4-trifluoromethyl-phenyl-acetic acid ethyl ester, benzeneacetic acid, 4-trifluoromethyl-, ethyl ester, ethyl4-trifluoromethyl phenylacetate, ethyl 4-trifluoromethylphenylacetate, ethyl 4-trifluoromethyl-phenyl acetate, ethyl 4-trifluoromethyl phenyl acetate | |
CCOC(=O)CC1=CC=C(C=C1)C(F)(F)F | |
232.20 | |
232.2 | |
Solid |
721-63-1 | |
C11H11F3O2 | |
250 mg | |
BDVKGYOFECBKDX-UHFFFAOYSA-N | |
ethyl 2-[4-(trifluoromethyl)phenyl]acetate | |
14134124 | |
95% |
Chemical Identifiers
721-63-1 | |
232.20 | |
BDVKGYOFECBKDX-UHFFFAOYSA-N | |
ethyl 2-[4-(trifluoromethyl)phenyl]acetate |
C11H11F3O2 | |
MFCD07779351 | |
14134124 | |
CCOC(=O)CC1=CC=C(C=C1)C(F)(F)F |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures