missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 5-fluoroindole-2-carboxylate, 98%, Thermo Scientific Chemicals

$114.82 - $373.27

Chemical Identifiers

CAS 348-36-7
Molecular Formula C11H10FNO2
Molecular Weight (g/mol) 207.20
MDL Number MFCD00152076
InChI Key VIKOQTQMWBKMNA-UHFFFAOYSA-N
Synonym ethyl 5-fluoroindole-2-carboxylate, 5-fluoroindole-2-carboxylic acid ethyl ester, 2-carbethoxy-5-fluoroindole, 1h-indole-2-carboxylic acid, 5-fluoro-, ethyl ester, 5-fluoro-1h-indole-2-carboxylic acid ethyl ester, ethyl5-fluoro-indole-2-carboxylate, pubchem7276, maybridge1_006190, acmc-1aex1, ksc573a9h
PubChem CID 2727899
IUPAC Name ethyl 5-fluoro-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=CC(F)=CC=C2N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1401003
View Documents
Thermo Scientific Chemicals
L1401003
1 g
Each for $114.82
 
AAL1401006
View Documents
Thermo Scientific Chemicals
L1401006
5 g
Each for $373.27
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

348-36-7
207.20
VIKOQTQMWBKMNA-UHFFFAOYSA-N
2727899
CCOC(=O)C1=CC2=CC(F)=CC=C2N1
C11H10FNO2
MFCD00152076
ethyl 5-fluoroindole-2-carboxylate, 5-fluoroindole-2-carboxylic acid ethyl ester, 2-carbethoxy-5-fluoroindole, 1h-indole-2-carboxylic acid, 5-fluoro-, ethyl ester, 5-fluoro-1h-indole-2-carboxylic acid ethyl ester, ethyl5-fluoro-indole-2-carboxylate, pubchem7276, maybridge1_006190, acmc-1aex1, ksc573a9h
ethyl 5-fluoro-1H-indole-2-carboxylate

Specifications

348-36-7
C11H10FNO2
1 g
ethyl 5-fluoroindole-2-carboxylate, 5-fluoroindole-2-carboxylic acid ethyl ester, 2-carbethoxy-5-fluoroindole, 1h-indole-2-carboxylic acid, 5-fluoro-, ethyl ester, 5-fluoro-1h-indole-2-carboxylic acid ethyl ester, ethyl5-fluoro-indole-2-carboxylate, pubchem7276, maybridge1_006190, acmc-1aex1, ksc573a9h
CCOC(=O)C1=CC2=CC(F)=CC=C2N1
207.20
207.21
Ethyl 5-fluoroindole-2-carboxylate
147°C to 152°C
MFCD00152076
170254
VIKOQTQMWBKMNA-UHFFFAOYSA-N
ethyl 5-fluoro-1H-indole-2-carboxylate
2727899
98%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only