missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 6-Aminopyridine-2-carboxylate 98.0+%, TCI America™
$192.00 - $579.71
Chemical Identifiers
| CAS | 69142-64-9 |
|---|---|
| Molecular Formula | C8H10N2O2 |
| Molecular Weight (g/mol) | 166.18 |
| MDL Number | MFCD01009842 |
| InChI Key | DTYDENRYZSEGNL-UHFFFAOYSA-N |
| Synonym | 6-Aminopyridine-2-carboxylic Acid Ethyl Ester, Ethyl 6-Aminopicolinate, 6-Aminopicolinic Acid Ethyl Ester |
| PubChem CID | 11959065 |
| IUPAC Name | ethyl 6-aminopyridine-2-carboxylate |
| SMILES | CCOC(=O)C1=NC(=CC=C1)N |
Chemical Identifiers
| 69142-64-9 | |
| 166.18 | |
| DTYDENRYZSEGNL-UHFFFAOYSA-N | |
| 11959065 | |
| CCOC(=O)C1=NC(=CC=C1)N |
| C8H10N2O2 | |
| MFCD01009842 | |
| 6-Aminopyridine-2-carboxylic Acid Ethyl Ester, Ethyl 6-Aminopicolinate, 6-Aminopicolinic Acid Ethyl Ester | |
| ethyl 6-aminopyridine-2-carboxylate |
Specifications
| 69142-64-9 | |
| Yellow | |
| MFCD01009842 | |
| 6-Aminopyridine-2-carboxylic Acid Ethyl Ester, Ethyl 6-Aminopicolinate, 6-Aminopicolinic Acid Ethyl Ester | |
| CCOC(=O)C1=NC(=CC=C1)N | |
| 166.18 | |
| 166.18 | |
| Crystalline Powder |
| 72°C | |
| C8H10N2O2 | |
| 1 g | |
| DTYDENRYZSEGNL-UHFFFAOYSA-N | |
| ethyl 6-aminopyridine-2-carboxylate | |
| 11959065 | |
| ≥98.0% (GC,T) | |
| Ethyl 6-Aminopyridine-2-carboxylate |
Safety and Handling
TSCA : No