missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 6-Methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine-5-carboxylate 98.0+%, TCI America™

Supplier:  TCI America E11311G

 View more versions of this product

Catalog No. E11311G


Explore available promotions

Description

Description

Specifications

Specifications

Ethyl 6-Methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine-5-carboxylate
208°C
C14H16N2O3
1 g
URMNHHAUVFEMIG-UHFFFAOYSA-N
ethyl 6-methyl-2-oxo-4-phenyl-3,4-dihydro-1H-pyrimidine-5-carboxylate
220272
≥98.0% (HPLC,N)
5395-36-8
White
MFCD00124778
6-Methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine-5-carboxylic Acid Ethyl Ester
CCOC(=O)C1=C(NC(=O)NC1C2=CC=CC=C2)C
260.293
260.29
Crystalline Powder

Chemical Identifiers

5395-36-8
260.293
URMNHHAUVFEMIG-UHFFFAOYSA-N
220272
CCOC(=O)C1=C(NC(=O)NC1C2=CC=CC=C2)C
C14H16N2O3
MFCD00124778
6-Methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine-5-carboxylic Acid Ethyl Ester
ethyl 6-methyl-2-oxo-4-phenyl-3,4-dihydro-1H-pyrimidine-5-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions