missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 6-Methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine-5-carboxylate 98.0+%, TCI America™
Supplier: TCI America E1131200MG
Specifications
Ethyl 6-Methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine-5-carboxylate | |
208°C | |
C14H16N2O3 | |
200 mg | |
URMNHHAUVFEMIG-UHFFFAOYSA-N | |
ethyl 6-methyl-2-oxo-4-phenyl-3,4-dihydro-1H-pyrimidine-5-carboxylate | |
220272 | |
≥98.0% (HPLC,N) |
5395-36-8 | |
White | |
MFCD00124778 | |
6-Methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine-5-carboxylic Acid Ethyl Ester | |
CCOC(=O)C1=C(NC(=O)NC1C2=CC=CC=C2)C | |
260.293 | |
260.29 | |
Crystalline Powder |
Chemical Identifiers
5395-36-8 | |
260.293 | |
URMNHHAUVFEMIG-UHFFFAOYSA-N | |
220272 | |
CCOC(=O)C1=C(NC(=O)NC1C2=CC=CC=C2)C |
C14H16N2O3 | |
MFCD00124778 | |
6-Methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine-5-carboxylic Acid Ethyl Ester | |
ethyl 6-methyl-2-oxo-4-phenyl-3,4-dihydro-1H-pyrimidine-5-carboxylate |
Safety and Handling
TSCA : No