missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl Acetate, ≥99.5%, ACS Reagent, Honeywell™ Riedel-de Haën™
ACS Reagent,≥99.5%
Supplier: Honeywell Chemicals 3199021L
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Ethyl Acetate | |
141-78-6 | |
77.5°C | |
2.2 to 11.5% (38°F) | |
801°F | |
C4H8O2 | |
MFCD00009171 | |
UN1173 | |
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester | |
CCOC(=O)C | |
88.106 | |
CHEBI:27750 | |
≥99.5% |
-84°C | |
0.902g/mL at 25°C | |
1 L | |
−3°C (26.6°F) | |
Glass Bottle | |
CH3COOC2H5 | |
73mmHg (20°C) | |
506104 | |
XEKOWRVHYACXOJ-UHFFFAOYSA-N | |
ethyl acetate | |
8857 | |
88.11g/mol | |
ACS Reagent |
Chemical Identifiers
141-78-6 | |
88.106 | |
XEKOWRVHYACXOJ-UHFFFAOYSA-N | |
8857 | |
ethyl acetate |
C4H8O2 | |
MFCD00009171 | |
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester | |
CHEBI:27750 | |
CCOC(=O)C |
Safety and Handling
P210-P261-P305 + P351 + P338
H225-H319-H336
DOTInformation : Class 3 - PG 2 - Ethyl acetate
EINECSNumber : 205-500-4
RTECSNumber : AH5425000