missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl acetate, +99.5%, for HPLC, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 268350040

 View more versions of this product

Catalog No. AC268350040



Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
Specifications

Specifications

Ethyl acetate
141-78-6
Colorless
0.9020g/mL
4 L
Liquid
0.0002 % max.
0.902
CH3CO2C2H5
Vapor Pressure: 103mbar at 20°C
0.03% Max.
15, 3811
Solubility in water: 80g/L (20°C). Other solubilities: soluble in benzene, acetone, chloroform, alcohol,, ether
CCOC(=O)C
88.106
CHEBI:27750
≥99.5%
-83.5°C
(1 cm cell vs HPLC−grade water), (1 cm cell vs HPLC−grade water) at 0nm, 0.005 max. at 280nm, 0.025 max. at 270nm, 0.12 max. at 260nm, 1 max. at 255nm
0.0015 meq/g max.
75.0°C to 78.0°C
−4°C
1.3710 to 1.3740
99.5+%
C4H8O2
MFCD00009171
0.45 mPa.s (20°C)
02, 125
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
8857
88.11
HPLC

Chemical Identifiers

141-78-6
88.106
XEKOWRVHYACXOJ-UHFFFAOYSA-N
8857
ethyl acetate
C4H8O2
MFCD00009171
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
CHEBI:27750
CCOC(=O)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 205-500-4

RTECSNumber : AH5425000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only