Learn More
Ethyl acetate, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
$90.02 - $291.12
Chemical Identifiers
CAS | 141-78-6 |
---|---|
Molecular Formula | C4H8O2 |
Molecular Weight (g/mol) | 88.106 |
MDL Number | MFCD00009171 |
InChI Key | XEKOWRVHYACXOJ-UHFFFAOYSA-N |
Synonym | ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester |
PubChem CID | 8857 |
ChEBI | CHEBI:27750 |
IUPAC Name | ethyl acetate |
SMILES | CCOC(=O)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC364351000
|
Thermo Scientific Chemicals
364351000 |
100 mL | AcroSeal™ Glass Bottle |
Each for $90.02
|
|
||||
AC364350010
|
Thermo Scientific Chemicals
364350010 |
1 L | AcroSeal™ Glass Bottle |
Each for $157.52
|
|
||||
AC364350025
|
Thermo Scientific Chemicals
364350025 |
2.5 L | AcroSeal™ Glass Bottle |
Each for $291.12
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
141-78-6 | |
88.106 | |
XEKOWRVHYACXOJ-UHFFFAOYSA-N | |
8857 | |
ethyl acetate |
C4H8O2 | |
MFCD00009171 | |
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester | |
CHEBI:27750 | |
CCOC(=O)C |
Specifications
-83.5°C | |
Colorless | |
75.0°C to 78.0°C | |
−4°C | |
AcroSeal™ Glass Bottle | |
1.3710 to 1.3740 | |
0.902 | |
CH3CO2C2H5 | |
0.45 mPa.s (20°C) | |
02, 125 | |
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester | |
XEKOWRVHYACXOJ-UHFFFAOYSA-N | |
ethyl acetate | |
8857 | |
88.11 | |
Water <50ppm |
141-78-6 | |
0.9020g/mL | |
100 mL | |
Authentic | |
Liquid | |
99.9% | |
C4H8O2 | |
MFCD00009171 | |
0.005% Max. (Coulometric) | |
15, 3811 | |
Solubility in water: 80g/L (20°C). Other solubilities: soluble in benzene, acetone, chloroform, alcohol,, ether | |
CCOC(=O)C | |
88.106 | |
CHEBI:27750 | |
99.9% | |
Ethyl acetate, Over Molecular Sieves |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 205-500-4
RTECSNumber : AH5425000
TSCA : TSCA
RUO – Research Use Only