missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acetate, 99+%, extra pure

CAS: 141-78-6 | C4H8O2 | 88.106 g/mol

$94.15 - $1229.11

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC149470010
SDS View Documents
Thermo Scientific Chemicals
149470010
1 L Glass Bottle
Each for $94.15
Case of 4 Each for $0.00
Only null left
 
AC149470025
SDS View Documents
Thermo Scientific Chemicals
149470025
2.5 L Glass Bottle
Each for $184.94
Only null left
 
AC149470050
SDS missing translation for 'viewDocuments'
Thermo Scientific Chemicals
149470050
5 L Plastic Drum
Each for $368.95
Only null left
 
AC149470250
SDS missing translation for 'viewDocuments'
Thermo Scientific Chemicals
149470250
25 L Metal Drum
Each for $1,229.11
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

141-78-6
88.106
XEKOWRVHYACXOJ-UHFFFAOYSA-N
8857
ethyl acetate
C4H8O2
MFCD00009171
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
CHEBI:27750
CCOC(=O)C

Specifications

-83.5°C
0.9020g/mL
1 L
Glass Bottle
1.3710 to 1.3740
99+%
C4H8O2
MFCD00009171
0.45 mPa.s (20°C)
02, 125
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
8857
88.11
Extra Pure
141-78-6
75.0°C to 78.0°C
−4°C
Liquid
0.005% max.
0.902
CH3CO2C2H5
Vapor Pressure: 103mbar at 20°C
0.05% max. (K.F.)
15, 3811
Solubility in water: 80g/L (20°C). Other solubilities: soluble in benzene, acetone, chloroform, alcohol,, ether
CCOC(=O)C
88.106
CHEBI:27750
≥99%
Ethyl acetate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
Documents

Documents

RUO – Research Use Only