missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Acetate, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.5%, Honeywell

Supplier:  Honeywell Chemicals LP1004

 View more versions of this product

Catalog No. 6000593


Explore available promotions

Description

Description

Specifications

Specifications

Ethyl Acetate
141-78-6
0.8945 g/cm3
4 x 4 L
Glass Bottle
C4H8O2
0.2% max.
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
8857
88.11
ACS Reagent
ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.5%
100.0000%
77°C
−4°C
by GC Analysis
UN1173
506104
870% in Water at 20°C
CCOC(=O)C
88.106
CHEBI:27750
>99.5%

Chemical Identifiers

141-78-6
88.106
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
CHEBI:27750
CCOC(=O)C
C4H8O2
XEKOWRVHYACXOJ-UHFFFAOYSA-N
8857
ethyl acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 547 days from date of manufacture

DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Ethyl acetate

SDS
Documents

Documents

Promotions

Promotions