missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl acetate, For pesticide residue analysis, Honeywell Riedel-de Haën™

CHROMASOLV™, for pesticide residue analysis

Supplier:  Honeywell-Chromasolv 3106325L

 View more versions of this product

Catalog No. 6000581


Explore available promotions

Description

Description

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent
Specifications

Specifications

Ethyl Acetate
0.902g/mL at 25°C
2.2 to 11.5% (38°F)
801°F
CH3COOC2H5
73mmHg (20°C)
506104
Soluble in water
CCOC(=O)C
88.106
CHEBI:27750
CHROMASOLV™
141-78-6
2.5 L
−3°C (26.6°F)
C4H8O2
MFCD00009171
1173
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
8857
88.11g/mol

Chemical Identifiers

141-78-6
88.106
XEKOWRVHYACXOJ-UHFFFAOYSA-N
8857
ethyl acetate
C4H8O2
MFCD00009171
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
CHEBI:27750
CCOC(=O)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P261-P305 + P351 + P338

H225-H319-H336

EINECSNumber : 205-500-4

RTECSNumber : AH5425000

SDS
Documents

Documents

Promotions

Promotions