missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl acetate, Environmental Grade, 99.5+%, Thermo Scientific Chemicals

Pesticide residue, environmental, and GC analysis

Supplier:  Thermo Scientific Chemicals 040977M1

 View more versions of this product

Catalog No. AA40977M1


Explore available promotions

Description

Description

Ethyl acetate is used primarily as a solvent and diluent. It is commonly used to clean the circuit boards, in some nail varnish removers and also in residue, environmental and GC analysis. It is used chiefly as a scent in the manufacture of perfumes. It is also used in flavorings, glues, in decaffeinating tea and coffee, and in cigarettes. The other minor applications of ethyl acetate include its applications in the textiles, dyestuffs, explosives, photographic films and plates, artificial leather.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl acetate
141-78-6
0.902 g/mL
4 x 1 L
Sweet
1.372
CH3CO2CH2CH3
UN1173
14,3757
Miscible with ethanol,acetone,diethyl ether and benzene.
CCOC(=O)C
88.106
CHEBI:27750
≥99.5%
-84°C
Colorless
77°C
-3°C (26°F)
Liquid
C4H8O2
MFCD00009171
506104
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
8857
88.11
Environmental

Chemical Identifiers

141-78-6
88.106
XEKOWRVHYACXOJ-UHFFFAOYSA-N
8857
ethyl acetate
C4H8O2
MFCD00009171
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
CHEBI:27750
CCOC(=O)C
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c

H225-H319-H336

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL ACETATE

EINECSNumber : 205-500-4

RTECSNumber : AH5425000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only