missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Acetate, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.5%, Honeywell Burdick & Jackson

$1279.08 - $9386.90

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
6000595
View Documents
Honeywell Chemicals
PP10019
19 L Metal Can
Each for $1,279.08
 
6000596
View Documents
Honeywell Chemicals
PP100204
204 L Metal Drum
Each for $9,386.90
 
Description

Description

Specifications

Chemical Identifiers

141-78-6
88.106
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
CHEBI:27750
CCOC(=O)C
C4H8O2
XEKOWRVHYACXOJ-UHFFFAOYSA-N
8857
ethyl acetate

Specifications

141-78-6
0.8945 g/cm3
19 L
Metal Can
UN1173
506104
870% in Water at 20°C
CCOC(=O)C
88.106
CHEBI:27750
>99.9%
Ethyl Acetate, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.5%
100.0000%
77°C
−4°C
C4H8O2
0.1% max.
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
8857
88.11
ACS Reagent
by GC Analysis
Videos
Safety and Handling

Safety and Handling

ShelfLife : 547 days from date of manufacture

DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Ethyl acetate

SDS
missing translation for 'documents'

missing translation for 'documents'