missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Acetate, Reagent, ACS, 99.5%, Spectrum™ Chemical

E1025, 141-78-6, C4H8O2

Supplier:  Spectrum Chemical Mfg Cor E10254LTGLCS4

 View more versions of this product

Catalog No. 18602506


Explore available promotions

Description

Description

Spectrum™ Chemical Ethyl Acetate, Reagent, ACS , is a colorless liquid that smells like pear drops. Abbreviated as EA, it can be found in glues, nail polish removers, cigarettes and other products. Primarily used as a diluent because of its low cost and toxicity, it is also used in laboratory settings for column chromatography and extractions. Prone to hydrolysis and transesterification, it is rarely used as a solvent. As an ACS grade Reagent, Spectrum Chemical manufactured Ethyl Acetate is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.
Specifications

Specifications

141-78-6
4 x 4 L
0.003%
UN1173
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
99.5%
100%
Amber Glass Bottle
C4H8O2
0.2%
CCOC(C)=O
88.11
ACS

Chemical Identifiers

141-78-6
88.11
ethyl acetate
C4H8O2
XEKOWRVHYACXOJ-UHFFFAOYSA-N
CCOC(C)=O
Videos
SDS
Documents

Documents

Promotions

Promotions