missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl acetate, Spectrophotometric Grade, 99.5+%, Thermo Scientific Chemicals

$104.25 - $309.38

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA39177K2
View Documents
Thermo Scientific Chemicals
039177K2
1 L
Each for $104.25
 
AA39177M1
View Documents
Thermo Scientific Chemicals
039177M1
4 x 1 L
Each for $309.38
 
AA39177K7
View Documents
Thermo Scientific Chemicals
039177K7
4 L
Each for $305.78
 
Description

Description

Ethyl acetate in its highly pure form is used as a solvent for spectrophotometric applications. It is most suited for HPLC instruments and spectrophotometers including UV-visible spectrophotometer, IR spectrometer and mass spectrometer. In addition, it can also be used as a solvent in column chromatography and solvent extraction. Also, it is used in glues, nail polish removers, decaffeinating tea and coffee, and cigarettes. It is present in confectioneries, perfumes, and fruits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

141-78-6
88.106
XEKOWRVHYACXOJ-UHFFFAOYSA-N
8857
ethyl acetate
C4H8O2
MFCD00009171
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
CHEBI:27750
CCOC(=O)C

Specifications

-84°C
Colorless
77°C
-3°C (26°F)
Liquid
C4H8O2
MFCD00009171
506104
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
8857
88.11
Spectrophotometric
141-78-6
0.902 g/mL
1 L
Sweet
1.372
CH3CO2CH2CH3
UN1173
14,3757
Miscible in ethanol,acetone,diethyl ether and benzene.
CCOC(=O)C
88.106
CHEBI:27750
≥99.5%
Ethyl acetate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c

H225-H319-H336

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL ACETATE

EINECSNumber : 205-500-4

RTECSNumber : AH5425000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only