missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl acetoacetate, 99+%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 220405000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl acetoacetate | |
141-97-9 | |
1.0300g/mL | |
70°C | |
99% min. (GC) | |
C6H10O3 | |
CH3COCH2CO2C2H5 | |
500 g | |
12,222 | |
15, 3812 | |
Solubility in water: 116g/L (20°C). Other solubilities: soluble in most organic solvent: diethyl ether,,benzene,ethanol,ehyl acetate,chloroform,,acetone | |
CCOC(=O)CC(=O)C | |
130.14 | |
1.696 mPa.s (20°C) | |
130.14 | |
Extra Pure |
p.a. | |
-45°C | |
180°C | |
Authentic | |
Glass bottle | |
1.4180 to 1.42 | |
MFCD00009199 | |
03, 632 | |
1.03 | |
ethyl acetoacetate, ethyl acetylacetate, ethyl 3-oxobutyrate, diacetic ether, butanoic acid, 3-oxo-, ethyl ester, ethyl acetylacetonate, acetoacetic acid, ethyl ester, 3-oxobutanoic acid ethyl ester, 3-oxo-butyric acid ethyl ester, active acetylacetate | |
XYIBRDXRRQCHLP-UHFFFAOYSA-N | |
ethyl 3-oxobutanoate | |
8868 | |
CHEBI:4893 | |
99+% | |
Liquid |
Chemical Identifiers
141-97-9 | |
130.14 | |
XYIBRDXRRQCHLP-UHFFFAOYSA-N | |
8868 | |
ethyl 3-oxobutanoate |
C6H10O3 | |
MFCD00009199 | |
ethyl acetoacetate, ethyl acetylacetate, ethyl 3-oxobutyrate, diacetic ether, butanoic acid, 3-oxo-, ethyl ester, ethyl acetylacetonate, acetoacetic acid, ethyl ester, 3-oxobutanoic acid ethyl ester, 3-oxo-butyric acid ethyl ester, active acetylacetate | |
CHEBI:4893 | |
CCOC(=O)CC(=O)C |
Safety and Handling
EINECSNumber : 205-516-1
RTECSNumber : AK5250000
TSCA : TSCA
RUO – Research Use Only