missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl acetoacetate, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1254430

 View more versions of this product

Catalog No. AAA1254430


Explore available promotions

Description

Description

Ethyl acetoacetate is used as an intermediate in organic synthesis and as a co-promoter for unsaturated polyester resins. It is widely used in the production of dyes, inks, perfumes, plastics and flavoring agents. It is an important starting material for the syntheses of alpha-substituted acetoacetic esters and cyclic compounds like pyrazole, pyrimidine and coumarin derivatives. It acts as an intermediate in the synthesis of vitamins and pharmaceuticals. It finds application as a formaldehyde scavenger.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl acetoacetate is used as an intermediate in organic synthesis and as a co-promoter for unsaturated polyester resins. It is widely used in the production of dyes, inks, perfumes, plastics and flavoring agents. It is an important starting material for the syntheses of alpha-substituted acetoacetic esters and cyclic compounds like pyrazole, pyrimidine and coumarin derivatives. It acts as an intermediate in the synthesis of vitamins and pharmaceuticals. It finds application as a formaldehyde scavenger.

Solubility
Miscible with water, benzene, chloroform, ether and acetone.

Notes
Incompatible with acids, bases, oxidizing agents, alkali metals and reducing agents.
Specifications

Specifications

Ethyl acetoacetate
-43°C
180°C to 181°C
≥99%
C6H10O3
MFCD00009199
385838
ethyl acetoacetate, ethyl acetylacetate, ethyl 3-oxobutyrate, diacetic ether, butanoic acid, 3-oxo-, ethyl ester, ethyl acetylacetonate, acetoacetic acid, ethyl ester, 3-oxobutanoic acid ethyl ester, 3-oxo-butyric acid ethyl ester, active acetylacetate
XYIBRDXRRQCHLP-UHFFFAOYSA-N
ethyl 3-oxobutanoate
8868
130.14
Liquid
141-97-9
1.025
84°C (183°F)
Fruit-like
1.419
250 g
14,3758
Miscible with water,benzene,chloroform,ether and acetone.
CCOC(=O)CC(=O)C
130.143
CHEBI:4893
≥99%

Chemical Identifiers

141-97-9
130.143
XYIBRDXRRQCHLP-UHFFFAOYSA-N
8868
ethyl 3-oxobutanoate
C6H10O3
MFCD00009199
ethyl acetoacetate, ethyl acetylacetate, ethyl 3-oxobutyrate, diacetic ether, butanoic acid, 3-oxo-, ethyl ester, ethyl acetylacetonate, acetoacetic acid, ethyl ester, 3-oxobutanoic acid ethyl ester, 3-oxo-butyric acid ethyl ester, active acetylacetate
CHEBI:4893
CCOC(=O)CC(=O)C
Safety and Handling

Safety and Handling

GHS H Statement
H319-H227
Causes serious eye irritation.
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 205-516-1

RTECSNumber : AK5250000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only