missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Acetoacetate 98.0+%, TCI America™

Supplier:  TCI America A0649500G

 View more versions of this product

Catalog No. A0649500G


Explore available promotions

Description

Description

Specifications

Specifications

Ethyl Acetoacetate
-43°C
184°C
MFCD00009199
ethyl acetoacetate, ethyl acetylacetate, ethyl 3-oxobutyrate, diacetic ether, butanoic acid, 3-oxo-, ethyl ester, ethyl acetylacetonate, acetoacetic acid, ethyl ester, 3-oxobutanoic acid ethyl ester, 3-oxo-butyric acid ethyl ester, active acetylacetate
CCOC(=O)CC(=O)C
130.143
CHEBI:4893
≥98.0% (GC)
141-97-9
Colorless
C6H10O3
500 g
XYIBRDXRRQCHLP-UHFFFAOYSA-N
ethyl 3-oxobutanoate
8868
130.14
Liquid

Chemical Identifiers

141-97-9
130.143
XYIBRDXRRQCHLP-UHFFFAOYSA-N
8868
ethyl 3-oxobutanoate
C6H10O3
MFCD00009199
ethyl acetoacetate, ethyl acetylacetate, ethyl 3-oxobutyrate, diacetic ether, butanoic acid, 3-oxo-, ethyl ester, ethyl acetylacetonate, acetoacetic acid, ethyl ester, 3-oxobutanoic acid ethyl ester, 3-oxo-butyric acid ethyl ester, active acetylacetate
CHEBI:4893
CCOC(=O)CC(=O)C
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1475&(2)-1505

RTECSNumber : AK5250000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions