Learn More
Ethyl acrylate, 99%, stab. with ca 20ppm 4-methoxyphenol, Thermo Scientific Chemicals
$63.74 - $83.63
Chemical Identifiers
CAS | 140-88-5 |
---|---|
Molecular Formula | C5H8O2 |
Molecular Weight (g/mol) | 100.12 |
MDL Number | MFCD00009188 |
InChI Key | JIGUQPWFLRLWPJ-UHFFFAOYSA-N |
Synonym | ethyl acrylate, acrylic acid ethyl ester, ethyl propenoate, 2-propenoic acid, ethyl ester, ethyl 2-propenoate, ethylacrylaat, ethylakrylat, etil acrilato, acrylic acid, ethyl ester, aethylacrylat |
PubChem CID | 8821 |
ChEBI | CHEBI:82327 |
IUPAC Name | ethyl prop-2-enoate |
SMILES | CCOC(=O)C=C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA15493AE
|
Thermo Scientific Chemicals
A15493AE |
100 mL |
Each for $63.74
|
|
|||||
AAA15493AP
|
Thermo Scientific Chemicals
A15493AP |
500 mL |
Each for $83.63
|
|
|||||
Description
Ethyl acrylate is used in the syntheses of various pharmaceutical intermediates, textiles and non-woven fibers. It is widely used in the production of polymers including resins, plastics, rubber and denture material. It is actively involved in the preparation of dimethylaminoethylacrylate (DMAEA) which is used as a monomer to make flocculants for water treatment. It plays a vital role in the manufacture of water-based latex paints and adhesives, textile and paper coatings and leather finish resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl acrylate is used in the syntheses of various pharmaceutical intermediates, textiles and non-woven fibers. It is widely used in the production of polymers including resins, plastics, rubber and denture material. It is actively involved in the preparation of dimethylaminoethylacrylate (DMAEA) which is used as a monomer to make flocculants for water treatment. It plays a vital role in the manufacture of water-based latex paints and adhesives, textile and paper coatings and leather finish resins.
Solubility
Miscible with ethanol, ethyl ether and chloroform. Silghtly miscible with water and dimethyl sulfoxide.
Notes
Store in cool place. Incompatible with strong oxidizing agents, strong acids, amines, peroxides, chlorosulfonic acid, moisture and strong alkalies.
Chemical Identifiers
140-88-5 | |
100.12 | |
JIGUQPWFLRLWPJ-UHFFFAOYSA-N | |
8821 | |
ethyl prop-2-enoate |
C5H8O2 | |
MFCD00009188 | |
ethyl acrylate, acrylic acid ethyl ester, ethyl propenoate, 2-propenoic acid, ethyl ester, ethyl 2-propenoate, ethylacrylaat, ethylakrylat, etil acrilato, acrylic acid, ethyl ester, aethylacrylat | |
CHEBI:82327 | |
CCOC(=O)C=C |
Specifications
140-88-5 | |
0.924 | |
−1°C (30°F) | |
C5H8O2 | |
CH2=CHCO2CH2CH3 | |
100 mL | |
773866 | |
ethyl acrylate, acrylic acid ethyl ester, ethyl propenoate, 2-propenoic acid, ethyl ester, ethyl 2-propenoate, ethylacrylaat, ethylakrylat, etil acrilato, acrylic acid, ethyl ester, aethylacrylat | |
JIGUQPWFLRLWPJ-UHFFFAOYSA-N | |
ethyl prop-2-enoate | |
8821 | |
100.12 | |
Ethyl acrylate, stabilized with ≈20ppm 4-methoxyphenol |
-71°C | |
99°C to 100°C | |
Pungent | |
1.406 | |
MFCD00009188 | |
UN1917 | |
14,3759 | |
Miscible with ethanol,ethyl ether and chloroform. Silghtly miscible with water and dimethyl sulfoxide. | |
CCOC(=O)C=C | |
100.12 | |
CHEBI:82327 | |
99% |
Safety and Handling
GHS H Statement
H225-H302-H312-H332-H315-H319-H317-H335
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P332+P313-P333+P313-P363
H225-H302+H312-H315-H317-H319-H331-H335-H351
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL ACRYLATE, STABILIZED
EINECSNumber : 205-438-8
RTECSNumber : AT0700000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only