missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Alcohol, Reagent, Denat., Spectro Grade, Contains 5% 2-PrOH and 5% MeOH, Thermo Scientific Chemicals

$102.31 - $180.12

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC224095000
SDS View Documents
Thermo Scientific Chemicals
224095000
500 mL Glass Bottle
Each for $102.31
 
AC224090010
SDS View Documents
Thermo Scientific Chemicals
224090010
1 L Glass Bottle
Each for $180.12
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for UV/VIS spectroscopy
Specifications

Chemical Identifiers

64-17-5
46.069
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
CHEBI:16236
CCO
C2H6O
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
702
ethanol

Specifications

64-17-5,67-56-1,67-63-0
1.59
Glass Bottle
0.0005% max.
C2H5OH
0.1% Max. (Coulometric)
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
CCO
46.069
CHEBI:16236
4.0 to 6.0% (2-propanol)
Ethanol
0.01 max. at 300nm, 0.04 max. at 250nm, 0.65 max. at 210nm, 1 max. at 205nm
500 mL
Liquid
C2H6O
Vapor Pressure: 59mbar at 20°C
14, 3761
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
702
46.06
Spectroscopy
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Manufacturing and Laboratory Use. Not for beverage use