Learn More
Ethyl alpha-bromophenylacetate, 97%, Thermo Scientific Chemicals
$124.75 - $1524.84
Chemical Identifiers
CAS | 2882-19-1 |
---|---|
Molecular Formula | C10H11BrO2 |
Molecular Weight (g/mol) | 243.10 |
MDL Number | MFCD00013536 |
InChI Key | BKTKLDMYHTUESO-UHFFFAOYNA-N |
Synonym | ethyl alpha-bromophenylacetate, ethyl bromophenylacetate, ethyl bromo phenyl acetate, ethyl-alpha-bromophenyl acetate, ethyl-.alpha.-bromophenyl acetate, benzeneacetic acid, .alpha.-bromo-, ethyl ester, bromo-phenyl-acetic acid ethyl ester, benzeneacetic acid, alpha-bromo-, ethyl ester, eyr&vo2, ethylbromo phenyl acetate |
PubChem CID | 97780 |
IUPAC Name | ethyl 2-bromo-2-phenylacetate |
SMILES | CCOC(=O)C(Br)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2111409
|
Thermo Scientific Chemicals
B2111409 |
10 g |
Each for $124.75
|
|
|||||
AAB2111418
|
Thermo Scientific Chemicals
B2111418 |
50 g |
Each for $421.92
|
|
|||||
AAB2111430
|
Thermo Scientific Chemicals
B2111430 |
250 g |
Each for $1,524.84
|
|
|||||
Description
α-Bromobenzeneacetic acid ethyl ester is used as an alkyl halide initiator in controlled/living radical polymerization of styrene and methyl methacrylate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsα-Bromobenzeneacetic acid ethyl ester is used as an alkyl halide initiator in controlled/living radical polymerization of styrene and methyl methacrylate.
Solubility
Not miscible or difficult to mix in water.
Notes
Moisture sensitive. Store away from oxidizing agents, air and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Chemical Identifiers
2882-19-1 | |
243.10 | |
BKTKLDMYHTUESO-UHFFFAOYNA-N | |
97780 | |
CCOC(=O)C(Br)C1=CC=CC=C1 |
C10H11BrO2 | |
MFCD00013536 | |
ethyl alpha-bromophenylacetate, ethyl bromophenylacetate, ethyl bromo phenyl acetate, ethyl-alpha-bromophenyl acetate, ethyl-.alpha.-bromophenyl acetate, benzeneacetic acid, .alpha.-bromo-, ethyl ester, bromo-phenyl-acetic acid ethyl ester, benzeneacetic acid, alpha-bromo-, ethyl ester, eyr&vo2, ethylbromo phenyl acetate | |
ethyl 2-bromo-2-phenylacetate |
Specifications
2882-19-1 | |
164°C (30 mmHg) | |
Pungent | |
1.5395 | |
10 g | |
Moisture sensitive | |
Not miscible or difficult to mix in water. | |
CCOC(=O)C(Br)C1=CC=CC=C1 | |
243.10 | |
243.1 | |
Ethyl alpha-bromophenylacetate |
1.389 | |
>110°C (230°F) | |
C10H11BrO2 | |
MFCD00013536 | |
UN3265 | |
ethyl alpha-bromophenylacetate, ethyl bromophenylacetate, ethyl bromo phenyl acetate, ethyl-alpha-bromophenyl acetate, ethyl-.alpha.-bromophenyl acetate, benzeneacetic acid, .alpha.-bromo-, ethyl ester, bromo-phenyl-acetic acid ethyl ester, benzeneacetic acid, alpha-bromo-, ethyl ester, eyr&vo2, ethylbromo phenyl acetate | |
BKTKLDMYHTUESO-UHFFFAOYNA-N | |
ethyl 2-bromo-2-phenylacetate | |
97780 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 220-735-2
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only