missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl alpha-Iodophenylacetate 98.0+%, TCI America™
$119.48 - $360.10
Chemical Identifiers
CAS | 78489-65-3 |
---|---|
Molecular Formula | C10H11IO2 |
Molecular Weight (g/mol) | 290.1 |
InChI Key | GFYUBPKXVMWINH-UHFFFAOYSA-N |
Synonym | alpha-Iodophenylacetic Acid Ethyl Ester, Ethyl 2-Iodo-2-phenylacetate, 2-Iodo-2-phenylacetic Acid Ethyl Ester |
PubChem CID | 13346227 |
IUPAC Name | ethyl 2-iodo-2-phenylacetate |
SMILES | CCOC(=O)C(C1=CC=CC=C1)I |
Chemical Identifiers
78489-65-3 | |
290.1 | |
alpha-Iodophenylacetic Acid Ethyl Ester, Ethyl 2-Iodo-2-phenylacetate, 2-Iodo-2-phenylacetic Acid Ethyl Ester | |
ethyl 2-iodo-2-phenylacetate |
C10H11IO2 | |
GFYUBPKXVMWINH-UHFFFAOYSA-N | |
13346227 | |
CCOC(=O)C(C1=CC=CC=C1)I |
Specifications
78489-65-3 | |
5 g | |
alpha-Iodophenylacetic Acid Ethyl Ester, Ethyl 2-Iodo-2-phenylacetate, 2-Iodo-2-phenylacetic Acid Ethyl Ester | |
CCOC(=O)C(C1=CC=CC=C1)I | |
290.1 | |
290.10 | |
Liquid |
C10H11IO2 | |
3265 | |
GFYUBPKXVMWINH-UHFFFAOYSA-N | |
ethyl 2-iodo-2-phenylacetate | |
13346227 | |
≥98.0% (HPLC) | |
Ethyl alpha-Iodophenylacetate |
Safety and Handling
TSCA : No
Recommended Storage : Freezer