Learn More
Ethyl benzimidate hydrochloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1768418
Description
Ethyl benzimidate hydrochloride is used in the preparation of (4R)-ethyl 2-phenyl-4,5-dihydrothiazole-4-carboxylate by reacting with (R)-ethyl cysteine hydrochloride. It also reacts with D-penicillamine methyl ester hydrochloride and triethylamine to prepare methyl-5,5- dimethyl-2-phenyl-2-thiazoline-4-carboxylate. Further, it is used as an intermediate for organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl benzimidate hydrochloride is used in the preparation of (4R)-ethyl 2-phenyl-4,5-dihydrothiazole-4-carboxylate by reacting with (R)-ethyl cysteine hydrochloride. It also reacts with D-penicillamine methyl ester hydrochloride and triethylamine to prepare methyl-5,5- dimethyl-2-phenyl-2-thiazoline-4-carboxylate. Further, it is used as an intermediate for organic synthesis.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Ethyl benzimidate hydrochloride | |
115°C to 117°C | |
MFCD00043246 | |
3697589 | |
ethyl benzimidate hydrochloride, benzenecarboximidic acid, ethyl ester, hydrochloride, ethyl benzenecarboximidate hydrochloride, benzimidic acid ethyl ester hcl, ethylbenzimidate hydrochloride, ethyl benzimidate hcl, ethyl benzylimidate hydrochloride, ethyl iminobenzoate hydrochloride, benzimido ethyl ether hydrochloride, benzimidoyl ethyl ether hydrochloride | |
MODZVIMSNXSQIH-UHFFFAOYSA-N | |
ethyl benzenecarboximidate;hydrochloride | |
79249 | |
97% |
5333-86-8 | |
C9H12ClNO | |
50 g | |
Hygroscopic | |
Soluble in water. | |
[H+].[Cl-].CCOC(=N)C1=CC=CC=C1 | |
185.65 | |
185.65 |
Chemical Identifiers
5333-86-8 | |
185.65 | |
MODZVIMSNXSQIH-UHFFFAOYSA-N | |
79249 | |
[H+].[Cl-].CCOC(=N)C1=CC=CC=C1 |
C9H12ClNO | |
MFCD00043246 | |
ethyl benzimidate hydrochloride, benzenecarboximidic acid, ethyl ester, hydrochloride, ethyl benzenecarboximidate hydrochloride, benzimidic acid ethyl ester hcl, ethylbenzimidate hydrochloride, ethyl benzimidate hcl, ethyl benzylimidate hydrochloride, ethyl iminobenzoate hydrochloride, benzimido ethyl ether hydrochloride, benzimidoyl ethyl ether hydrochloride | |
ethyl benzenecarboximidate;hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 226-248-1
TSCA : Yes
Recommended Storage : Store under Nitrogen
RUO – Research Use Only