missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Benzoate, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 118095000

 View more versions of this product

Catalog No. AC118095000

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl benzoate
93-89-0
-34.0°C
1.0450g/mL
88°C
99% min. (GC)
C9H10O2
C6H5CO2C2H5
500 g
1.045
benzoic acid, ethyl ester, benzoic ether, ethyl benzenecarboxylate, ethylbenzoate, benzoyl ethyl ether, benzoic acid ethyl ester, ethylester kyseliny benzoove, natural, unii-j115brj15h, fema no. 2422
MTZQAGJQAFMTAQ-UHFFFAOYSA-N
ethyl benzoate
7165
99+%
99+%
1mg KOH/g max.
Colorless to Yellow
212.0°C
Authentic
Glass bottle
1.5040 to 1.5060
MFCD00009109
09, IV, 285
15, 3821
Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and chloroform
CCOC(=O)C1=CC=CC=C1
150.18
150.18
Liquid

Chemical Identifiers

93-89-0
150.18
MTZQAGJQAFMTAQ-UHFFFAOYSA-N
7165
CCOC(=O)C1=CC=CC=C1
C9H10O2
MFCD00009109
benzoic acid, ethyl ester, benzoic ether, ethyl benzenecarboxylate, ethylbenzoate, benzoyl ethyl ether, benzoic acid ethyl ester, ethylester kyseliny benzoove, natural, unii-j115brj15h, fema no. 2422
ethyl benzoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 202-284-3

RTECSNumber : DH0200000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only