missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl bromoacetate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1044836

 View more versions of this product

Catalog No. AAA1044836


Explore available promotions

Description

Description

Ethyl bromoacetate is widely used as an alkylating reagent involved in the Reformatsky reaction to prepare the beta-hydroxy esters by reacting with carbonyl compounds. It is also used for the syntheses of witting reagent, artificial diethylstilbestrol antigen, 3-phenyl-1-naphthol and steroidal thiazolidinone derivatives. It finds applications in the preparation of reversibly photoresponsive coumarin-stabilized polymeric nanoparticles in an aqueous medium which act as a detectable drug carrier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl bromoacetate is widely used as an alkylating reagent involved in the Reformatsky reaction to prepare the beta-hydroxy esters by reacting with carbonyl compounds. It is also used for the syntheses of witting reagent, artificial diethylstilbestrol antigen, 3-phenyl-1-naphthol and steroidal thiazolidinone derivatives. It finds applications in the preparation of reversibly photoresponsive coumarin-stabilized polymeric nanoparticles in an aqueous medium which act as a detectable drug carrier.

Solubility
Miscible with ethanol, acetone, benzene and ethyl ether. Immiscible with water.

Notes
It is a lachrymator. Incompatible with strong oxidizing agents.
Specifications

Specifications

Ethyl bromoacetate
-38°C
159°C
98%
1.451
MFCD00000191
UN1603
Miscible with ethanol,acetone,benzene and ethyl ether. Immiscible with water.
CCOC(=O)CBr
167.002
167
105-36-2
1.506
47°C (117°F)
C4H7BrO2
BrCH2CO2CH2CH3
500 g
ethyl bromoacetate, antol, ethyl bromacetate, bromoacetic acid ethyl ester, ethyl monobromoacetate, acetic acid, bromo-, ethyl ester, ethoxycarbonylmethyl bromide, bromoacetic acid, ethyl ester, ethyl alpha-bromoacetate, ethyl bromo acetate
PQJJJMRNHATNKG-UHFFFAOYSA-N
ethyl 2-bromoacetate
7748
98%

Chemical Identifiers

105-36-2
167.002
PQJJJMRNHATNKG-UHFFFAOYSA-N
7748
CCOC(=O)CBr
C4H7BrO2
MFCD00000191
ethyl bromoacetate, antol, ethyl bromacetate, bromoacetic acid ethyl ester, ethyl monobromoacetate, acetic acid, bromo-, ethyl ester, ethoxycarbonylmethyl bromide, bromoacetic acid, ethyl ester, ethyl alpha-bromoacetate, ethyl bromo acetate
ethyl 2-bromoacetate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H226
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapor.

P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P310-P330-P363-P370+P378q-P501c

H226-H300+H310+H330

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ETHYL BROMOACETATE

EINECSNumber : 203-290-9

RTECSNumber : AF6000000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only