missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl bromoacetate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 158591000

 View more versions of this product

Catalog No. AC158591000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl bromoacetate
105-36-2
1.5060g/mL
47°C
97.5% min. (GC)
C4H7BrO2
BrCH2CO2C2H5
100 g
02,192; 13,60
ethyl bromoacetate, antol, ethyl bromacetate, bromoacetic acid ethyl ester, ethyl monobromoacetate, acetic acid, bromo-, ethyl ester, ethoxycarbonylmethyl bromide, bromoacetic acid, ethyl ester, ethyl alpha-bromoacetate, ethyl bromo acetate
PQJJJMRNHATNKG-UHFFFAOYSA-N
ethyl 2-bromoacetate
7748
98%
98%
-38.0°C
159.0°C
Authentic
Glass bottle
1.4500 to 1.4520
MFCD00000191
02, 214
1.506
Solubility in water: insoluble in water. Other solubilities: soluble in alcohol, benzene and ether
CCOC(=O)CBr
167
167
Liquid

Chemical Identifiers

105-36-2
PQJJJMRNHATNKG-UHFFFAOYSA-N
7748
CCOC(=O)CBr
MFCD00000191
ethyl bromoacetate, antol, ethyl bromacetate, bromoacetic acid ethyl ester, ethyl monobromoacetate, acetic acid, bromo-, ethyl ester, ethoxycarbonylmethyl bromide, bromoacetic acid, ethyl ester, ethyl alpha-bromoacetate, ethyl bromo acetate
ethyl 2-bromoacetate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if inhaled.
Fatal if swallowed.
Fatal in contact with skin.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Keep away from heat/sparks/open flames/hot

GHS Signal Word: Danger

EINECSNumber : 203-290-9

RTECSNumber : AF6000000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only