Learn More
Ethyl bromoacetate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 158595000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl bromoacetate | |
105-36-2 | |
1.5060g/mL | |
47°C | |
97.5% min. (GC) | |
C4H7BrO2 | |
BrCH2CO2C2H5 | |
500 g | |
02,192; 13,60 | |
ethyl bromoacetate, antol, ethyl bromacetate, bromoacetic acid ethyl ester, ethyl monobromoacetate, acetic acid, bromo-, ethyl ester, ethoxycarbonylmethyl bromide, bromoacetic acid, ethyl ester, ethyl alpha-bromoacetate, ethyl bromo acetate | |
PQJJJMRNHATNKG-UHFFFAOYSA-N | |
ethyl 2-bromoacetate | |
7748 | |
98% |
98% | |
-38.0°C | |
159.0°C | |
Authentic | |
Glass bottle | |
1.4500 to 1.4520 | |
MFCD00000191 | |
02, 214 | |
1.506 | |
Solubility in water: insoluble in water. Other solubilities: soluble in alcohol, benzene and ether | |
CCOC(=O)CBr | |
167 | |
167 | |
Liquid |
Chemical Identifiers
105-36-2 | |
PQJJJMRNHATNKG-UHFFFAOYSA-N | |
7748 | |
CCOC(=O)CBr |
MFCD00000191 | |
ethyl bromoacetate, antol, ethyl bromacetate, bromoacetic acid ethyl ester, ethyl monobromoacetate, acetic acid, bromo-, ethyl ester, ethoxycarbonylmethyl bromide, bromoacetic acid, ethyl ester, ethyl alpha-bromoacetate, ethyl bromo acetate | |
ethyl 2-bromoacetate |
Safety and Handling
GHS H Statement
Fatal if inhaled.
Fatal if swallowed.
Fatal in contact with skin.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Keep away from heat/sparks/open flames/hot
GHS Signal Word: Danger
EINECSNumber : 203-290-9
RTECSNumber : AF6000000
TSCA : TSCA
RUO – Research Use Only