Learn More
Ethyl bromopyruvate, 80-85%
CAS: 70-23-5 | C5H7BrO3 | 195.01 g/mol
$194.85 - $194.85
Chemical Identifiers
| CAS | 70-23-5 |
|---|---|
| Molecular Formula | C5H7BrO3 |
| Molecular Weight (g/mol) | 195.01 |
| MDL Number | MFCD00000204 |
| InChI Key | VICYTAYPKBLQFB-UHFFFAOYSA-N |
| Synonym | ethyl bromopyruvate, ethyl 3-bromopyruvate, propanoic acid, 3-bromo-2-oxo-, ethyl ester, bromopyruvic acid ethyl ester, ethyl bromo pyruvate, ethyl 3-bromo-2-oxopropionate, pyruvic acid, bromo-, ethyl ester, .beta.-bromopyruvic acid ethyl ester, 3-bromo-2-oxo-propionic acid ethyl ester, beta-bromopyruvic acid ethyl ester |
| PubChem CID | 66144 |
| IUPAC Name | ethyl 3-bromo-2-oxopropanoate |
| SMILES | CCOC(=O)C(=O)CBr |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC118150250
|
Thermo Scientific Chemicals
118150250 |
25 mL | Glass bottle |
Each for $194.85
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 70-23-5 | |
| 195.01 | |
| VICYTAYPKBLQFB-UHFFFAOYSA-N | |
| 66144 | |
| CCOC(=O)C(=O)CBr |
| C5H7BrO3 | |
| MFCD00000204 | |
| ethyl bromopyruvate, ethyl 3-bromopyruvate, propanoic acid, 3-bromo-2-oxo-, ethyl ester, bromopyruvic acid ethyl ester, ethyl bromo pyruvate, ethyl 3-bromo-2-oxopropionate, pyruvic acid, bromo-, ethyl ester, .beta.-bromopyruvic acid ethyl ester, 3-bromo-2-oxo-propionic acid ethyl ester, beta-bromopyruvic acid ethyl ester | |
| ethyl 3-bromo-2-oxopropanoate |
Specifications
| 70-23-5 | |
| 1.5500g/mL | |
| 98°C | |
| 80% min. (GC) | |
| C5H7BrO3 | |
| BrCH2COCO2C2H5 | |
| 25 mL | |
| 1.55 | |
| VICYTAYPKBLQFB-UHFFFAOYSA-N | |
| ethyl 3-bromo-2-oxopropanoate | |
| 66144 | |
| 80-85% | |
| Ethyl bromopyruvate |
| Pink-Red to Yellow | |
| 100.0°C to 102.0°C (15.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4675 to 1.4765 | |
| MFCD00000204 | |
| 03, II, 409 | |
| ethyl bromopyruvate, ethyl 3-bromopyruvate, propanoic acid, 3-bromo-2-oxo-, ethyl ester, bromopyruvic acid ethyl ester, ethyl bromo pyruvate, ethyl 3-bromo-2-oxopropionate, pyruvic acid, bromo-, ethyl ester, .beta.-bromopyruvic acid ethyl ester, 3-bromo-2-oxo-propionic acid ethyl ester, beta-bromopyruvic acid ethyl ester | |
| CCOC(=O)C(=O)CBr | |
| 195.01 | |
| 195.01 | |
| Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 200-729-6
RUO – Research Use Only