Learn More
Ethyl butyrate, 99%
CAS: 105-54-4 | C6H12O2 | 116.16 g/mol
$35.76 - $135.22
Chemical Identifiers
| CAS | 105-54-4 |
|---|---|
| Molecular Formula | C6H12O2 |
| Molecular Weight (g/mol) | 116.16 |
| MDL Number | MFCD00009394 |
| InChI Key | OBNCKNCVKJNDBV-UHFFFAOYSA-N |
| Synonym | ethyl butyrate, butyric acid ethyl ester, ethyl n-butyrate, butanoic acid, ethyl ester, butyric ester, butyric ether, butyric acid, ethyl ester, ethyl n-butanoate, butanoic acid ethyl ester, fema number 2427 |
| PubChem CID | 7762 |
| IUPAC Name | ethyl butanoate |
| SMILES | CCCC(=O)OCC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC118180050
|
Thermo Scientific Chemicals
118180050 |
5 mL | Glass bottle |
Each for $35.76
|
|
||||
|
AC118182500
|
Thermo Scientific Chemicals
118182500 |
250 mL | Glass bottle |
Each for $57.48
|
|
||||
|
AC118180010
|
Thermo Scientific Chemicals
118180010 |
1 L | Glass bottle |
Each for $135.22
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 105-54-4 | |
| 116.16 | |
| OBNCKNCVKJNDBV-UHFFFAOYSA-N | |
| 7762 | |
| CCCC(=O)OCC |
| C6H12O2 | |
| MFCD00009394 | |
| ethyl butyrate, butyric acid ethyl ester, ethyl n-butyrate, butanoic acid, ethyl ester, butyric ester, butyric ether, butyric acid, ethyl ester, ethyl n-butanoate, butanoic acid ethyl ester, fema number 2427 | |
| ethyl butanoate |
Specifications
| 105-54-4 | |
| -93.3°C | |
| 0.8700g/mL | |
| 26°C | |
| 98.5% min. (GC) | |
| C6H12O2 | |
| CH3CH2CH2CO2C2H5 | |
| 5 mL | |
| 0.87 | |
| ethyl butyrate, butyric acid ethyl ester, ethyl n-butyrate, butanoic acid, ethyl ester, butyric ester, butyric ether, butyric acid, ethyl ester, ethyl n-butanoate, butanoic acid ethyl ester, fema number 2427 | |
| OBNCKNCVKJNDBV-UHFFFAOYSA-N | |
| ethyl butanoate | |
| 7762 | |
| 99% | |
| Ethyl butyrate, 99% |
| 1mg KOH/g max. | |
| Colorless | |
| 120.0°C | |
| Authentic | |
| Glass bottle | |
| 1.3910 to 1.3930 | |
| MFCD00009394 | |
| 02, 270 | |
| 15, 3830 | |
| Solubility in water: practically insoluble | |
| CCCC(=O)OCC | |
| 116.16 | |
| 116.16 | |
| Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
GHS P Statement
Wear eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a well-ventilated place.
Keep cool.
GHS Signal Word: Warning
EINECSNumber : 203-306-4
RTECSNumber : ET1660000
TSCA : TSCA
RUO – Research Use Only