missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl chloroacetate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1555436

 View more versions of this product

Catalog No. AAA1555436


Explore available promotions

Description

Description

Ethyl chloroacetate is a reagent used in the preparation of 5 member heterocycles. It is used as pharmaceutical and organic intermediate. It is used as a solvent for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl chloroacetate is a reagent used in the preparation of 5 member heterocycles. It is used as pharmaceutical and organic intermediate. It is used as a solvent for organic synthesis.

Solubility
Soluble in water 20 g/L (20°C).

Notes
Incompatible with acids, bases, oxidizing agents, reducing agents.
Specifications

Specifications

Ethyl chloroacetate
-26°C
142°C to 144°C
Pungent
1.421
MFCD00000932
UN1181
14,3783
Soluble in water 20g/L (20°C).
CCOC(=O)CCl
122.548
122.55
105-39-5
1.159
54°C (149°F)
C4H7ClO2
ClCH2CO2CH2CH3
500 g
506455
ethyl chloroacetate, ethyl chloracetate, ethyl monochloroacetate, acetic acid, chloro-, ethyl ester, chloroacetic acid ethyl ester, ethyl monochloracetate, ethyl chloroethanoate, ethyl alpha-chloroacetate, 2-chloroacetic acid ethyl ester, unii-6h07teh4tf
VEUUMBGHMNQHGO-UHFFFAOYSA-N
ethyl 2-chloroacetate
7751
99%

Chemical Identifiers

105-39-5
122.548
VEUUMBGHMNQHGO-UHFFFAOYSA-N
7751
CCOC(=O)CCl
C4H7ClO2
MFCD00000932
ethyl chloroacetate, ethyl chloracetate, ethyl monochloroacetate, acetic acid, chloro-, ethyl ester, chloroacetic acid ethyl ester, ethyl monochloracetate, ethyl chloroethanoate, ethyl alpha-chloroacetate, 2-chloroacetic acid ethyl ester, unii-6h07teh4tf
ethyl 2-chloroacetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H226
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Flammable liquid and vapour.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P312-P330-P333+P313-P363-P370+P378q-P501c

H226-H300-H311+H331-H315-H317-H318

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ETHYL CHLOROACETATE

EINECSNumber : 203-294-0

RTECSNumber : AF9110000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only