missing translation for 'onlineSavingsMsg'
Learn More

Ethyl cis-9-Hexadecenoate 95.0+%, TCI America™

$294.52 - $294.52

Chemical Identifiers

CAS 56219-10-4
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.468
MDL Number MFCD00056314
InChI Key JELGPLUONQGOHF-KTKRTIGZSA-N
Synonym Ethyl Palmitoleate, cis-9-Hexadecenoic Acid Ethyl Ester, Palmitoleic Acid Ethyl Ester
PubChem CID 6436624
ChEBI CHEBI:84934
IUPAC Name ethyl (Z)-hexadec-9-enoate
SMILES CCCCCCC=CCCCCCCCC(=O)OCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H0501100MG
View Documents
TCI America
H0501100MG
100 mg N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

56219-10-4
282.468
JELGPLUONQGOHF-KTKRTIGZSA-N
6436624
ethyl (Z)-hexadec-9-enoate
C18H34O2
MFCD00056314
Ethyl Palmitoleate, cis-9-Hexadecenoic Acid Ethyl Ester, Palmitoleic Acid Ethyl Ester
CHEBI:84934
CCCCCCC=CCCCCCCCC(=O)OCC

Specifications

56219-10-4
135°C
MFCD00056314
Ethyl Palmitoleate, cis-9-Hexadecenoic Acid Ethyl Ester, Palmitoleic Acid Ethyl Ester
CCCCCCC=CCCCCCCCC(=O)OCC
282.468
CHEBI:84934
≥95.0% (GC)
Ethyl cis-9-Hexadecenoate
Colorless
C18H34O2
100 mg
JELGPLUONQGOHF-KTKRTIGZSA-N
ethyl (Z)-hexadec-9-enoate
6436624
282.47
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
missing translation for 'documents'

missing translation for 'documents'