missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl coumarin-3-carboxylate, 98%, Thermo Scientific Chemicals
$45.48 - $139.70
Chemical Identifiers
CAS | 1846-76-0 |
---|---|
Molecular Formula | C12H10O4 |
Molecular Weight (g/mol) | 218.208 |
MDL Number | MFCD00016964 |
InChI Key | XKHPEMKBJGUYCM-UHFFFAOYSA-N |
Synonym | ethyl coumarin-3-carboxylate, ethyl 2-oxo-2h-chromene-3-carboxylate, 3-ethoxycarbonylcoumarin, ethyl 3-coumarincarboxylate, 3-carbethoxycoumarin, ethylcoumarin-3-carboxylate, coumarin-3-carboxylic acid ethyl ester, 1,2-benzopyran-2-one-3-carboxylic acid, ethyl ester, 2h-1-benzopyran-3-carboxylic acid, 2-oxo-, ethyl ester, 2h-1-benzopyran-3-carboxylicacid, 2-oxo-, ethyl ester |
PubChem CID | 15800 |
IUPAC Name | ethyl 2-oxochromene-3-carboxylate |
SMILES | CCOC(=O)C1=CC2=CC=CC=C2OC1=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1255303
|
Thermo Scientific Chemicals
A1255303 |
1 g |
Each for $45.48
|
|
|||||
AAA1255306
|
Thermo Scientific Chemicals
A1255306 |
5 g |
Each for $139.70
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1846-76-0 | |
218.208 | |
XKHPEMKBJGUYCM-UHFFFAOYSA-N | |
15800 | |
CCOC(=O)C1=CC2=CC=CC=C2OC1=O |
C12H10O4 | |
MFCD00016964 | |
ethyl coumarin-3-carboxylate, ethyl 2-oxo-2h-chromene-3-carboxylate, 3-ethoxycarbonylcoumarin, ethyl 3-coumarincarboxylate, 3-carbethoxycoumarin, ethylcoumarin-3-carboxylate, coumarin-3-carboxylic acid ethyl ester, 1,2-benzopyran-2-one-3-carboxylic acid, ethyl ester, 2h-1-benzopyran-3-carboxylic acid, 2-oxo-, ethyl ester, 2h-1-benzopyran-3-carboxylicacid, 2-oxo-, ethyl ester | |
ethyl 2-oxochromene-3-carboxylate |
Specifications
1846-76-0 | |
C12H10O4 | |
1 g | |
ethyl coumarin-3-carboxylate, ethyl 2-oxo-2h-chromene-3-carboxylate, 3-ethoxycarbonylcoumarin, ethyl 3-coumarincarboxylate, 3-carbethoxycoumarin, ethylcoumarin-3-carboxylate, coumarin-3-carboxylic acid ethyl ester, 1,2-benzopyran-2-one-3-carboxylic acid, ethyl ester, 2h-1-benzopyran-3-carboxylic acid, 2-oxo-, ethyl ester, 2h-1-benzopyran-3-carboxylicacid, 2-oxo-, ethyl ester | |
CCOC(=O)C1=CC2=CC=CC=C2OC1=O | |
218.208 | |
218.21 | |
Ethyl coumarin-3-carboxylate |
92°C to 95°C | |
MFCD00016964 | |
200147 | |
XKHPEMKBJGUYCM-UHFFFAOYSA-N | |
ethyl 2-oxochromene-3-carboxylate | |
15800 | |
98% |
Safety and Handling
RTECSNumber : DJ2501000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only