missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl Ether, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, contains BHT, >98.5%, Honeywell
Supplier: Honeywell Chemicals LP10819
Specifications
Ethyl Ether | |
60-29-7,60-29-7 | |
0.713 g/cm3 | |
19 L | |
Metal Can | |
C4H10O | |
UN1155 | |
1696894 | |
689% in Water at 20°C | |
CCOCC | |
74.12 | |
CHEBI:35702 | |
>98% |
ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, contains BHT, >98.5% | |
100.0000%,≤100.0000% | |
35°C | |
−45°C | |
by GC Analysis | |
MFCD00011646 | |
0.03% max. | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
ethoxyethane | |
3283 | |
74.12 | |
ACS Reagent |
Chemical Identifiers
60-29-7 | |
74.12 | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
3283 | |
ethoxyethane |
C4H10O | |
MFCD00011646 | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
CHEBI:35702 | |
CCOCC |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: I; Proper Shipping Name: Diethyl ether