missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Ether, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, contains BHT, >98.5%, Honeywell

Supplier:  Honeywell Inc LP1084

 View more versions of this product

Catalog No. 6000533


Explore available promotions

Description

Description

Specifications

Specifications

Ethyl Ether
60-29-7,60-29-7
0.713 g/cm3
4 x 4 L
Glass Bottle
C4H10O
UN1155
1696894
689% in Water at 20°C
CCOCC
74.12
CHEBI:35702
>98%
ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, contains BHT, >98.5%
100.0000%,≤100.0000%
35°C
−45°C
by GC Analysis
MFCD00011646
0.03% max.
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
RTZKZFJDLAIYFH-UHFFFAOYSA-N
ethoxyethane
3283
74.12
ACS Reagent

Chemical Identifiers

60-29-7
74.12
RTZKZFJDLAIYFH-UHFFFAOYSA-N
3283
ethoxyethane
C4H10O
MFCD00011646
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CHEBI:35702
CCOCC
Videos
Safety and Handling

Safety and Handling

ShelfLife : 547 days from date of manufacture

DOTInformation : Transport Hazard Class: 3; Packing Group:3: I; Proper Shipping Name: Diethyl ether

SDS
Documents

Documents

Promotions

Promotions