missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
![](http://beta-static.fishersci.ca/images/Solvent.gif)
![](http://beta-static.fishersci.ca/images/Red.gif)
Ethyl Ether Anhydrous (Certified ACS), Fisher Chemical™
Meet or exceeds the latest ACS specifications. Actual lot of analysis on label
$328.92 - $2,243.25
Chemical Identifiers
CAS | 60-29-7 |
---|---|
Molecular Formula | C4H10O |
Molecular Weight (g/mol) | 74.12 |
MDL Number | MFCD00011646 |
InChI Key | RTZKZFJDLAIYFH-UHFFFAOYSA-N |
Synonym | diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether |
PubChem CID | 3283 |
ChEBI | CHEBI:35702 |
IUPAC Name | ethoxyethane |
SMILES | CCOCC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
E138-500
|
Fisher Chemical
BPE138500 |
500 mL | Aluminum Can |
N/A
|
|
||||
E138-1
|
Fisher Chemical
FLE1381 |
1 L | Aluminum Can |
N/A
|
|
||||
E138-4
|
Fisher Chemical
BPE1384 |
4 L | Aluminum Can |
N/A
|
|
||||
E138-20
|
Fisher Chemical
FLE13820 |
20 L | Steel Pail |
N/A
|
|
||||
Description
Stabilized with BHT as preservative. Meet or exceeds the latest ACS specifications. Actual lot of analysis on label
Chemical Identifiers
60-29-7 | |
74.12 | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
3283 | |
ethoxyethane |
C4H10O | |
MFCD00011646 | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
CHEBI:35702 | |
CCOCC |
Specifications
-116°C | |
Colorless | |
34.6°C | |
-45°C (-111°F) | |
Aluminum Can | |
0.001% max. | |
0.0002mEq/g max. | |
C2H5OC2H5 | |
587mbar at 20°C | |
1155 | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
CCOCC | |
74.12 | |
CHEBI:35702 | |
≥99% | |
Ethyl Ether Anhydrous |
60-29-7 | |
0.714g/cm³ | |
500 mL | |
Pass Test | |
Liquid | |
≥99 % | |
C4H10O | |
MFCD00011646 | |
0.2448 cP at 20°C | |
0.01% max. | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
ethoxyethane | |
3283 | |
74.12 | |
Certified ACS Reagent |
Safety and Handling
![](http://beta-static.fishersci.ca/images/Solvent.gif)
![](http://beta-static.fishersci.ca/images/Red.gif)
DANGER!
Emergency Overview
Extremely flammable liquid and vapor. Vapor may cause flash fire. Harmful if swallowed. Irritating to eyes and skin. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. May cause irritation of respiratory tract. Repeated exposure may cause skin dryness or cracking. Hygroscopic. Use personal protective equipment. Keep away from open flames
DOTInformation : DOT Class 3, : Flammable Liquid