missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing


Ethyl Ether, Anhydrous (Stabilized/HPLC), Fisher Chemical
Supplier: Thermo Fisher Scientific Formerly Fairlawn FLE1984
Specifications
Ethyl Ether Anhydrous | |
Stabilized | |
0.01 AU max. at 350nm, 0.02 AU max. at 280nm, 0.07 AU max. at 254nm, 1.00 AU max. at 218nm | |
0.714g/cm³ | |
4 L | |
Pass Test | |
Liquid | |
1.3490 to 1.3520 (at 25°C) | |
99.0% min by GC | |
C4H10O | |
MFCD00011646 | |
0.2448 cP at 20°C | |
0.01% max. | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
ethoxyethane | |
3283 | |
74.12 | |
HPLC |
-116°C | |
60-29-7 | |
Colorless | |
34.6°C | |
-45°C (-111°F) | |
Tin Can | |
Mobile phase for HPLC applications | |
5ppm max. | |
0.0002mEq/g max. | |
C2H5OC2H5 | |
587mbar at 20°C | |
1155 | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
CCOCC | |
74.12 | |
CHEBI:35702 | |
≥99% |
Chemical Identifiers
60-29-7 | |
74.12 | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
3283 | |
ethoxyethane |
C4H10O | |
MFCD00011646 | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
CHEBI:35702 | |
CCOCC |
Safety and Handling


DOTInformation : Class 3: Flammable Liquid