Learn More
Ethyl Ether (Laboratory), Fisher Chemical™
Supplier: Thermo Fisher Scientific Formerly Fairlawn FLE1344
Specifications
Ethyl Ether | |
60-29-7 | |
0.714g/cm³ | |
4 L | |
Aluminum Can | |
C4H10O | |
MFCD00011646 | |
0.2448 cP at 20°C | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
ethoxyethane | |
3283 | |
74.12 |
-116°C | |
Colorless | |
34.6°C | |
Pass Test | |
Liquid | |
C2H5OC2H5 | |
587mbar at 20°C | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
CCOCC | |
74.12 | |
CHEBI:35702 | |
Laboratory |
Chemical Identifiers
60-29-7 | |
74.12 | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
3283 | |
ethoxyethane |
C4H10O | |
MFCD00011646 | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
CHEBI:35702 | |
CCOCC |
Safety and Handling
DANGER!
Emergency Overview
Extremely flammable liquid and vapor. Vapor may cause flash fire. Harmful if swallowed. Irritating to eyes and skin. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. May cause irritation of respiratory tract. Repeated exposure may cause skin dryness or cracking. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Handle under an inert atmosphere. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention.
NFPA
Health:1
Flammability:4
Instability:1
DOTInformation : DOT Class 3, : Flammable Liquid