missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Ether, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, contains BHT, >99.9%, Honeywell™ Burdick & Jackson
$2862.00 - $2862.00
Chemical Identifiers
| CAS | 60-29-7 |
|---|---|
| Molecular Formula | C4H10O |
| Molecular Weight (g/mol) | 74.12 |
| MDL Number | MFCD00011646 |
| InChI Key | RTZKZFJDLAIYFH-UHFFFAOYSA-N |
| Synonym | diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether |
| PubChem CID | 3283 |
| ChEBI | CHEBI:35702 |
| IUPAC Name | ethoxyethane |
| SMILES | CCOCC |
Chemical Identifiers
| 60-29-7 | |
| 74.12 | |
| RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
| 3283 | |
| ethoxyethane |
| C4H10O | |
| MFCD00011646 | |
| diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
| CHEBI:35702 | |
| CCOCC |
Specifications
| 60-29-7,60-29-7 | |
| 0.713 g/cm3 | |
| 19 L | |
| Metal Can | |
| MFCD00011646 | |
| 0.03% max. | |
| diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
| RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
| ethoxyethane | |
| 3283 | |
| 74.12 | |
| ACS Reagent | |
| by GC Analysis |
| 100.0000%,≤100.0000% | |
| 35°C | |
| −45°C | |
| C4H10O | |
| UN1155 | |
| 1696894 | |
| 689% in Water at 20°C | |
| CCOCC | |
| 74.12 | |
| CHEBI:35702 | |
| >99.9% | |
| Ethyl Ether, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, contains BHT, >99.9% |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: I; Proper Shipping Name: Diethyl ether