missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl glutaryl chloride, 97%, Thermo Scientific Chemicals

$105.18 - $392.63

Chemical Identifiers

CAS 5205-39-0
Molecular Formula C7H11ClO3
Molecular Weight (g/mol) 178.612
MDL Number MFCD00013659
InChI Key KKJAQUGGQMCNJY-UHFFFAOYSA-N
Synonym ethyl 4-chloroformyl butyrate, ethyl 5-chloro-5-oxovalerate, glutaric acid monoethyl ester chloride, pentanoic acid, 5-chloro-5-oxo-, ethyl ester, ethyl 4-chlorocarbonyl butanoate, pubchem16647, ksc939c3b, gamma-carbethoxybutyryl chloride, glutaric acid monoethylester chloride, 5-chloro-5-oxovaleric acid ethyl ester
PubChem CID 78881
IUPAC Name ethyl 5-chloro-5-oxopentanoate
SMILES CCOC(=O)CCCC(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59443AA
View Documents
Thermo Scientific Chemicals
H59443AA
5 mL
Each for $105.18
 
AAH59443AC
View Documents
Thermo Scientific Chemicals
H59443AC
25 mL
Each for $392.63
 
Description

Description

Ethyl glutaryl chloride is a main cationic monomer, it can bring quaternary ammonium salt

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl glutaryl chloride is a main cationic monomer, it can bring quaternary ammonium salt

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

5205-39-0
178.612
KKJAQUGGQMCNJY-UHFFFAOYSA-N
78881
CCOC(=O)CCCC(=O)Cl
C7H11ClO3
MFCD00013659
ethyl 4-chloroformyl butyrate, ethyl 5-chloro-5-oxovalerate, glutaric acid monoethyl ester chloride, pentanoic acid, 5-chloro-5-oxo-, ethyl ester, ethyl 4-chlorocarbonyl butanoate, pubchem16647, ksc939c3b, gamma-carbethoxybutyryl chloride, glutaric acid monoethylester chloride, 5-chloro-5-oxovaleric acid ethyl ester
ethyl 5-chloro-5-oxopentanoate

Specifications

5205-39-0
68°C to 70°C (0.07 mmHg)
1.443
MFCD00013659
UN3265
ethyl 4-chloroformyl butyrate, ethyl 5-chloro-5-oxovalerate, glutaric acid monoethyl ester chloride, pentanoic acid, 5-chloro-5-oxo-, ethyl ester, ethyl 4-chlorocarbonyl butanoate, pubchem16647, ksc939c3b, gamma-carbethoxybutyryl chloride, glutaric acid monoethylester chloride, 5-chloro-5-oxovaleric acid ethyl ester
KKJAQUGGQMCNJY-UHFFFAOYSA-N
ethyl 5-chloro-5-oxopentanoate
78881
97%
Ethyl glutaryl chloride
1.139
C7H11ClO3
ClCO(CH2)3COOC2H5
5 mL
1099817
Soluble in water.
CCOC(=O)CCCC(=O)Cl
178.612
178.61
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 226-000-2

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only