missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl imidazole-2-carboxylate, 97%, Thermo Scientific Chemicals

$147.92 - $529.59

Chemical Identifiers

CAS 33543-78-1
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.142
MDL Number MFCD03426031
InChI Key UHYNYIGCGVDBTC-UHFFFAOYSA-N
Synonym ethyl imidazole-2-carboxylate, 1h-imidazole-2-carboxylic acid, ethyl ester, ethyl1h-imidazole-2-carboxylate, 1h-imidazole-2-carboxylic acid ethyl ester, ethyl 2-imidazolecarboxylate, imidazole-2-carboxylic acid ethyl ester, 2-ethoxycarbonyl-1h-imidazole, ethylimidazole-2-carboxylate, ethoxyformyl imidazole, zlchem 847
PubChem CID 549404
IUPAC Name ethyl 1H-imidazole-2-carboxylate
SMILES CCOC(=O)C1=NC=CN1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1940503
View Documents
Thermo Scientific Chemicals
L1940503
1 g N/A
 
AAL1940506
View Documents
Thermo Scientific Chemicals
L1940506
5 g N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

33543-78-1
140.142
UHYNYIGCGVDBTC-UHFFFAOYSA-N
549404
CCOC(=O)C1=NC=CN1
C6H8N2O2
MFCD03426031
ethyl imidazole-2-carboxylate, 1h-imidazole-2-carboxylic acid, ethyl ester, ethyl1h-imidazole-2-carboxylate, 1h-imidazole-2-carboxylic acid ethyl ester, ethyl 2-imidazolecarboxylate, imidazole-2-carboxylic acid ethyl ester, 2-ethoxycarbonyl-1h-imidazole, ethylimidazole-2-carboxylate, ethoxyformyl imidazole, zlchem 847
ethyl 1H-imidazole-2-carboxylate

Specifications

33543-78-1
48°C to 51°C (0.3 mmHg)
MFCD03426031
115685
UHYNYIGCGVDBTC-UHFFFAOYSA-N
ethyl 1H-imidazole-2-carboxylate
549404
97%
176°C to 178°C
C6H8N2O2
1 g
ethyl imidazole-2-carboxylate, 1h-imidazole-2-carboxylic acid, ethyl ester, ethyl1h-imidazole-2-carboxylate, 1h-imidazole-2-carboxylic acid ethyl ester, ethyl 2-imidazolecarboxylate, imidazole-2-carboxylic acid ethyl ester, 2-ethoxycarbonyl-1h-imidazole, ethylimidazole-2-carboxylate, ethoxyformyl imidazole, zlchem 847
CCOC(=O)C1=NC=CN1
140.142
140.14
Ethyl imidazole-2-carboxylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only