Learn More
Ethyl imidazole-4-carboxylate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6409506
Description
A ring-substituted-1H-imidazole-4-carboxylic acid derivative, that is shown to act as a new class of anti-tuberculosis agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA ring-substituted-1H-imidazole-4-carboxylic acid derivative, that is shown to act as a new class of anti-tuberculosis agents.
Solubility
Soluble in dimethyl sulfoxide and methanol. Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications
Ethyl imidazole-4-carboxylate | |
156°C to 160°C | |
MFCD02646639,MFCD07363812 | |
606937 | |
Soluble in dimethyl sulfoxide and methanol. Slightly soluble in water. | |
CCOC(=O)C1=CN=CN1 | |
140.14 | |
140.14 | |
Solid |
23785-21-9 | |
C6H8N2O2 | |
5 g | |
ethyl 1h-imidazole-4-carboxylate, ethyl imidazole-4-carboxylate, ethyl 4-imidazolecarboxylate, ethyl 4 5-imidazolecarboxylate, 4-carbethoxyimidazole, 1h-imidazole-4-carboxylic acid ethyl ester, nic7lr0zon, unii-nic7lr0zon, imidazole-4-carboxylic acid ethyl ester, 1h-imidazole-4-carboxylic acid, ethyl ester | |
KLWYPRNPRNPORS-UHFFFAOYSA-N | |
ethyl 1H-imidazole-5-carboxylate | |
99170 | |
98% |
Chemical Identifiers
23785-21-9 | |
140.14 | |
KLWYPRNPRNPORS-UHFFFAOYSA-N | |
99170 | |
CCOC(=O)C1=CN=CN1 |
C6H8N2O2 | |
MFCD02646639,MFCD07363812 | |
ethyl 1h-imidazole-4-carboxylate, ethyl imidazole-4-carboxylate, ethyl 4-imidazolecarboxylate, ethyl 4 5-imidazolecarboxylate, 4-carbethoxyimidazole, 1h-imidazole-4-carboxylic acid ethyl ester, nic7lr0zon, unii-nic7lr0zon, imidazole-4-carboxylic acid ethyl ester, 1h-imidazole-4-carboxylic acid, ethyl ester | |
ethyl 1H-imidazole-5-carboxylate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only