missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl isobutyrylacetate, 95%, Thermo Scientific Chemicals
$664.65 - $664.65
Chemical Identifiers
CAS | 7152-15-0 |
---|---|
Molecular Formula | C8H14O3 |
Molecular Weight (g/mol) | 158.2 |
MDL Number | MFCD00009198 |
InChI Key | XCLDSQRVMMXWMS-UHFFFAOYSA-N |
Synonym | ethyl isobutyrylacetate, 4-methyl-3-oxo-pentanoic acid ethyl ester, pentanoic acid, 4-methyl-3-oxo-, ethyl ester, ethyl isobutyroylacetate, valeric acid, 4-methyl-3-oxo-, ethyl ester, .gamma.,.gamma.-dimethylacetoacetic acid ethyl ester, beta-keto-gamma-methylvaleric acid ethyl ester, ethyl alpha-isobutyroylacetate, ethylisobutyrylacetate, ethyl iso-butyrylacetate |
PubChem CID | 81583 |
IUPAC Name | ethyl 4-methyl-3-oxopentanoate |
SMILES | CCOC(=O)CC(=O)C(C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC383440500
|
Thermo Scientific Chemicals
383440500 |
50 g | Glass bottle |
Each for $664.65
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7152-15-0 | |
158.2 | |
XCLDSQRVMMXWMS-UHFFFAOYSA-N | |
81583 | |
CCOC(=O)CC(=O)C(C)C |
C8H14O3 | |
MFCD00009198 | |
ethyl isobutyrylacetate, 4-methyl-3-oxo-pentanoic acid ethyl ester, pentanoic acid, 4-methyl-3-oxo-, ethyl ester, ethyl isobutyroylacetate, valeric acid, 4-methyl-3-oxo-, ethyl ester, .gamma.,.gamma.-dimethylacetoacetic acid ethyl ester, beta-keto-gamma-methylvaleric acid ethyl ester, ethyl alpha-isobutyroylacetate, ethylisobutyrylacetate, ethyl iso-butyrylacetate | |
ethyl 4-methyl-3-oxopentanoate |
Specifications
7152-15-0 | |
Colorless to Yellow | |
173.0°C | |
Authentic | |
Glass bottle | |
1.4255 to 1.4275 | |
MFCD00009198 | |
0.981 | |
XCLDSQRVMMXWMS-UHFFFAOYSA-N | |
ethyl 4-methyl-3-oxopentanoate | |
81583 | |
95% | |
Ethyl isobutyrylacetate |
-9.0°C | |
0.9810g/mL | |
44°C | |
94% min. (Mixture of tautomers) (GC) | |
C8H14O3 | |
(CH3)2CHCOCH2CO2C2H5 | |
50 g | |
ethyl isobutyrylacetate, 4-methyl-3-oxo-pentanoic acid ethyl ester, pentanoic acid, 4-methyl-3-oxo-, ethyl ester, ethyl isobutyroylacetate, valeric acid, 4-methyl-3-oxo-, ethyl ester, .gamma.,.gamma.-dimethylacetoacetic acid ethyl ester, beta-keto-gamma-methylvaleric acid ethyl ester, ethyl alpha-isobutyroylacetate, ethylisobutyrylacetate, ethyl iso-butyrylacetate | |
CCOC(=O)CC(=O)C(C)C | |
158.2 | |
158.2 | |
Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Warning
EINECSNumber : 230-491-9
RUO – Research Use Only