missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl isovalerate, 98%, Thermo Scientific Chemicals

$45.64 - $378.56

Chemical Identifiers

CAS 108-64-5
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00009203
InChI Key PPXUHEORWJQRHJ-UHFFFAOYSA-N
Synonym ethyl isovalerate, ethyl 3-methylbutyrate, ethylisovalerate, ethyl isopentanoate, isovaleric acid, ethyl ester, butanoic acid, 3-methyl-, ethyl ester, ethyl beta-methylbutyrate, ethyl isovalerianate, 3-methylbutyric acid ethyl ester, 3-methylbutanoic acid ethyl ester
PubChem CID 7945
ChEBI CHEBI:31571
IUPAC Name ethyl 3-methylbutanoate
SMILES CCOC(=O)CC(C)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA10292AE
View Documents
Thermo Scientific Chemicals
A10292AE
100 mL
Each for $45.64
 
AAA10292AP
View Documents
Thermo Scientific Chemicals
A10292AP
500 mL
Each for $96.79
 
AAA102920F
View Documents
Thermo Scientific Chemicals
A102920F
2500 mL
Each for $378.56
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

108-64-5
130.187
PPXUHEORWJQRHJ-UHFFFAOYSA-N
7945
ethyl 3-methylbutanoate
C7H14O2
MFCD00009203
ethyl isovalerate, ethyl 3-methylbutyrate, ethylisovalerate, ethyl isopentanoate, isovaleric acid, ethyl ester, butanoic acid, 3-methyl-, ethyl ester, ethyl beta-methylbutyrate, ethyl isovalerianate, 3-methylbutyric acid ethyl ester, 3-methylbutanoic acid ethyl ester
CHEBI:31571
CCOC(=O)CC(C)C

Specifications

108-64-5
0.866
26°C (78°F)
C7H14O2
MFCD00009203
UN3272
14,3816
PPXUHEORWJQRHJ-UHFFFAOYSA-N
ethyl 3-methylbutanoate
7945
130.19
Ethyl isovalerate
-99°C
131°C to 133°C
Fruit-like
1.396
100 mL
1744677
ethyl isovalerate, ethyl 3-methylbutyrate, ethylisovalerate, ethyl isopentanoate, isovaleric acid, ethyl ester, butanoic acid, 3-methyl-, ethyl ester, ethyl beta-methylbutyrate, ethyl isovalerianate, 3-methylbutyric acid ethyl ester, 3-methylbutanoic acid ethyl ester
CCOC(=O)CC(C)C
130.187
CHEBI:31571
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c

H226-H315-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

EINECSNumber : 203-602-3

RTECSNumber : NY1504000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only