missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl isovalerate, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 150480050

Catalog No. AC150480050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Ethyl isovalerate
108-64-5
131°C to 133°C
Authentic
Glass bottle
1.3950 to 1.398
MFCD00009203
02, 312
15, 3871
Solubility in water: < 1%. Other solubilities: miscible with alcohol,benzene and ether
CCOC(=O)CC(C)C
130.19
CHEBI:31571
99%
99%
0.8630g/mL
26°C
98.5% min. (GC)
C7H14O2
(CH3)2CHCH2CO2C2H5
5mL
0.863
ethyl isovalerate, ethyl 3-methylbutyrate, ethylisovalerate, ethyl isopentanoate, isovaleric acid, ethyl ester, butanoic acid, 3-methyl-, ethyl ester, ethyl beta-methylbutyrate, ethyl isovalerianate, 3-methylbutyric acid ethyl ester, 3-methylbutanoic acid ethyl ester
PPXUHEORWJQRHJ-UHFFFAOYSA-N
ethyl 3-methylbutanoate
7945
130.19
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-602-3

RTECSNumber : NY1504000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only