missing translation for 'onlineSavingsMsg'
Learn More

Ethyl L-lactate, 99%

CAS: 687-47-8 | C5H10O3 | 118.132 g/mol

$183.59 - $711.42

Chemical Identifiers

CAS 687-47-8
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00065359
InChI Key LZCLXQDLBQLTDK-BYPYZUCNSA-N
Synonym ethyl l-lactate, ethyl 2s-2-hydroxypropanoate, l-ethyl lactate, l---ethyl lactate, ethyl l--lactate, ethyl s-2-hydroxypropionate, ethyl l---lactate, s-ethyl lactate, propanoic acid, 2-hydroxy-, ethyl ester, 2s
PubChem CID 92831
ChEBI CHEBI:78322
IUPAC Name ethyl (2S)-2-hydroxypropanoate
SMILES CCOC(=O)C(C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1090036
View Documents
Thermo Scientific Chemicals
A1090036
500 g
Each for $183.59
Only null left
 
AAA109000E
View Documents
Thermo Scientific Chemicals
A109000E
2500 g
Each for $711.42
Only null left
 
Description

Description

Ethyl L-lactate is used in the preparation of food additives, aryl aldimines, fragrances and in pharmaceutical preparations. It can be considered as a green solvent in chemical industries due to its biodegradable nature. As a solvent, it used in the production of nitrocellulose, cellulose acetate and cellulose ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl L-lactate is used in the preparation of food additives, aryl aldimines, fragrances and in pharmaceutical preparations. It can be considered as a green solvent in chemical industries due to its biodegradable nature. As a solvent, it used in the production of nitrocellulose, cellulose acetate and cellulose ethers.

Solubility
Miscible with water and chloroform.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

687-47-8
118.132
LZCLXQDLBQLTDK-BYPYZUCNSA-N
92831
ethyl (2S)-2-hydroxypropanoate
C5H10O3
MFCD00065359
ethyl l-lactate, ethyl 2s-2-hydroxypropanoate, l-ethyl lactate, l---ethyl lactate, ethyl l--lactate, ethyl s-2-hydroxypropionate, ethyl l---lactate, s-ethyl lactate, propanoic acid, 2-hydroxy-, ethyl ester, 2s
CHEBI:78322
CCOC(=O)C(C)O

Specifications

687-47-8
1.036 g/mL
46°C (114°F)
C5H10O3
MFCD00065359
UN1192
14,3817
Miscible with water and chloroform.
CCOC(=O)C(C)O
118.132
CHEBI:78322
99%
°C to 25°C
152°C to 155°C
99%
1.413
500 g
1720839
ethyl l-lactate, ethyl 2s-2-hydroxypropanoate, l-ethyl lactate, l---ethyl lactate, ethyl l--lactate, ethyl s-2-hydroxypropionate, ethyl l---lactate, s-ethyl lactate, propanoic acid, 2-hydroxy-, ethyl ester, 2s
LZCLXQDLBQLTDK-BYPYZUCNSA-N
ethyl (2S)-2-hydroxypropanoate
92831
118.13
Ethyl L-lactate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H226-H335

P210-P233-P240-P243-P261-P270-P271-P280-P301+P312-P304+P340-P305+P351+P338-P310-P312-P330-P374-P380-P501c

H226-H302-H318-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHYL LACTATE

EINECSNumber : 211-694-1

RTECSNumber : OD5070000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only