missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl L-(-)-O-Tosyllactate 98.0+%, TCI America™
$52.19 - $460.43
Chemical Identifiers
| CAS | 57057-80-4 |
|---|---|
| Molecular Formula | C12H16O5S |
| Molecular Weight (g/mol) | 272.315 |
| MDL Number | MFCD06809725 |
| InChI Key | SNLMUZGICZJWKN-JTQLQIEISA-N |
| Synonym | Ethyl L-(-)-O-(p-Toluenesulfonyl)lactate, L-(-)-O-(p-Toluenesulfonyl)lactic Acid Ethyl Ester, L-(-)-O-Tosyllactic Acid Ethyl Ester |
| PubChem CID | 11043905 |
| IUPAC Name | ethyl (2S)-2-(4-methylphenyl)sulfonyloxypropanoate |
| SMILES | CCOC(=O)C(C)OS(=O)(=O)C1=CC=C(C=C1)C |
Chemical Identifiers
| 57057-80-4 | |
| 272.315 | |
| SNLMUZGICZJWKN-JTQLQIEISA-N | |
| 11043905 | |
| CCOC(=O)C(C)OS(=O)(=O)C1=CC=C(C=C1)C |
| C12H16O5S | |
| MFCD06809725 | |
| Ethyl L-(-)-O-(p-Toluenesulfonyl)lactate, L-(-)-O-(p-Toluenesulfonyl)lactic Acid Ethyl Ester, L-(-)-O-Tosyllactic Acid Ethyl Ester | |
| ethyl (2S)-2-(4-methylphenyl)sulfonyloxypropanoate |
Specifications
| 57057-80-4 | |
| C12H16O5S | |
| 1 g | |
| SNLMUZGICZJWKN-JTQLQIEISA-N | |
| ethyl (2S)-2-(4-methylphenyl)sulfonyloxypropanoate | |
| 11043905 | |
| ≥98.0% (T) | |
| Ethyl L-(-)-O-Tosyllactate |
| White-Yellow | |
| MFCD06809725 | |
| Ethyl L-(-)-O-(p-Toluenesulfonyl)lactate, L-(-)-O-(p-Toluenesulfonyl)lactic Acid Ethyl Ester, L-(-)-O-Tosyllactic Acid Ethyl Ester | |
| CCOC(=O)C(C)OS(=O)(=O)C1=CC=C(C=C1)C | |
| 272.315 | |
| 272.32 | |
| Crystals |
Safety and Handling
TSCA : No