missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Lactate 98.0+%, TCI America™

$46.60 - $106.25

Chemical Identifiers

CAS 97-64-3
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00065359
InChI Key LZCLXQDLBQLTDK-UHFFFAOYSA-N
Synonym ethyl lactate, actylol, acytol, solactol, lactic acid, ethyl ester, propanoic acid, 2-hydroxy-, ethyl ester, lactate d'ethyle, 2-hydroxypropanoic acid ethyl ester, ethyl 2-hydroxypropionate, ethyl alpha-hydroxypropionate
PubChem CID 7344
ChEBI CHEBI:78321
IUPAC Name ethyl 2-hydroxypropanoate
SMILES CCOC(=O)C(C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
L000325ML
View Documents
TCI America
L000325ML
25 mL
Each for $46.60
Only null left
 
L0003500ML
View Documents
TCI America
L0003500ML
500 mL
Each for $106.25
Only null left
 
Description

Description

Specifications

Chemical Identifiers

97-64-3
118.132
LZCLXQDLBQLTDK-UHFFFAOYSA-N
7344
ethyl 2-hydroxypropanoate
C5H10O3
MFCD00065359
ethyl lactate, actylol, acytol, solactol, lactic acid, ethyl ester, propanoic acid, 2-hydroxy-, ethyl ester, lactate d'ethyle, 2-hydroxypropanoic acid ethyl ester, ethyl 2-hydroxypropionate, ethyl alpha-hydroxypropionate
CHEBI:78321
CCOC(=O)C(C)O

Specifications

97-64-3
Colorless
C5H10O3
25 mL
ethyl lactate, actylol, acytol, solactol, lactic acid, ethyl ester, propanoic acid, 2-hydroxy-, ethyl ester, lactate d'ethyle, 2-hydroxypropanoic acid ethyl ester, ethyl 2-hydroxypropionate, ethyl alpha-hydroxypropionate
CCOC(=O)C(C)O
118.132
CHEBI:78321
≥98.0% (GC)
Ethyl Lactate
-25°C
154°C
MFCD00065359
1192
LZCLXQDLBQLTDK-UHFFFAOYSA-N
ethyl 2-hydroxypropanoate
7344
118.13
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1371

RTECSNumber : OD5075000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'