missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl maltol, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 450801000

Catalog No. AC450801000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Ethyl maltol
289.0°C to 290.0°C
97.5% min. (HPLC)
100g
3-benzyloxy cyclobutanecarboxylic acid, 3-benzyloxy-cyclobutanecarboxylic acid, 3-benzyloxy cyclobutane-1-carboxylic acid, 3-benzyloxycyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 3-phenylmethoxy, trans-3-benzyloxycyclobutanecarboxylic acid, cis-3-benzyloxy cyclobutanecarboxylic acid, trans-3-benzyloxy cyclobutanecarboxylic acid, cis-3-benzyloxycyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 3-phenylmethoxy-, trans
C1C(CC1OCC2=CC=CC=C2)C(=O)O
140.14
140.14
4940-11-8
Authentic
C7H8O3
15,3879
YNNOFVDQHAHVFG-UHFFFAOYSA-N
3-phenylmethoxycyclobutane-1-carboxylic acid
12815111
98%
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 225-582-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only