missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl methanesulfonate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1293809

 View more versions of this product

Catalog No. AAA1293809

Explore available promotions

Description

Description

Ethyl methanesulfonate is used as mutagen for both mammalian and plant cells. It finds application as a model alkylating agent in the study of deoxyribonucleic acid (DNA) repair processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl methanesulfonate is used as mutagen for both mammalian and plant cells. It finds application as a model alkylating agent in the study of deoxyribonucleic acid (DNA) repair processes.

Solubility
Miscible with water.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS available online at www.alfa.com.Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

Ethyl Methanesulfonate
1.170
100°C (212°F)
C3H8O3S
MFCD00007559
UN2810
Moisture sensitive
ethyl mesylate, ethyl methanesulphonate, half-myleran, methanesulfonic acid, ethyl ester, methanesulfonic acid ethyl ester, methylsulfonic acid, ethyl ester, rcra waste number u119, ethyl methansulphonate, ethyl ester of methylsulfonic acid, ethyl ester of methanesulfonic acid
PLUBXMRUUVWRLT-UHFFFAOYSA-N
ethyl methanesulfonate
6113
124.16
62-50-0
85°C to 86°C (10 mm)
99%
1.418
10 g
773969
14,3827
Miscible with water.
CCOS(=O)(=O)C
124.154
CHEBI:23994
99%

Chemical Identifiers

62-50-0
124.154
PLUBXMRUUVWRLT-UHFFFAOYSA-N
6113
ethyl methanesulfonate
C3H8O3S
MFCD00007559
ethyl mesylate, ethyl methanesulphonate, half-myleran, methanesulfonic acid, ethyl ester, methanesulfonic acid ethyl ester, methylsulfonic acid, ethyl ester, rcra waste number u119, ethyl methansulphonate, ethyl ester of methylsulfonic acid, ethyl ester of methanesulfonic acid
CHEBI:23994
CCOS(=O)(=O)C
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H340-H351-H361-H302

P201-P202-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c

H302-H315-H319-H335-H340-H351

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 200-536-7

RTECSNumber : PB2100000

TSCA : Yes

storageNote1 : Moisture Sensitive

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only