Learn More
Ethyl methyl sulfide, 97%, Thermo Scientific Chemicals
$94.22 - $261.14
Chemical Identifiers
CAS | 624-89-5 |
---|---|
Molecular Formula | C3H8S |
Molecular Weight (g/mol) | 76.16 |
MDL Number | MFCD00009268 |
InChI Key | WXEHBUMAEPOYKP-UHFFFAOYSA-N |
Synonym | ethyl methyl sulfide, methylthioethane, ethane, methylthio, methyl ethyl sulfide, ethyl methyl sulphide, methyl ethyl sulphide, sulfide, ethyl methyl, unii-mr1gzs62tm, methylthio ethane, methylsulfanyl ethane |
PubChem CID | 12230 |
IUPAC Name | methylsulfanylethane |
SMILES | CCSC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0285914
|
Thermo Scientific Chemicals
L0285914 |
25 g |
Each for $94.22
|
|
|||||
AAL0285922
|
Thermo Scientific Chemicals
L0285922 |
100 g |
Each for $261.14
|
|
|||||
Description
Ethyl Methyl Sulfide is used as a corrosion inhibitor for zinc metal which finds wide applications like electrode material in batteries, sacrificial anodes and metallic coatings. It is used in mixtures designed to odorize natural gas and is made to by-pass injection system.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl Methyl Sulfide is used as a corrosion inhibitor for zinc metal which finds wide applications like electrode material in batteries, sacrificial anodes and metallic coatings. It is used in mixtures designed to odorize natural gas and is made to by-pass injection system.
Solubility
Miscible with alcohols and oils. Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
624-89-5 | |
76.16 | |
WXEHBUMAEPOYKP-UHFFFAOYSA-N | |
12230 | |
CCSC |
C3H8S | |
MFCD00009268 | |
ethyl methyl sulfide, methylthioethane, ethane, methylthio, methyl ethyl sulfide, ethyl methyl sulphide, methyl ethyl sulphide, sulfide, ethyl methyl, unii-mr1gzs62tm, methylthio ethane, methylsulfanyl ethane | |
methylsulfanylethane |
Specifications
624-89-5 | |
0.841 | |
−15°C (5°F) | |
C3H8S | |
CH3CH2SCH3 | |
25 g | |
1696871 | |
Miscible with alcohols and oils. Immiscible with water. | |
CCSC | |
76.16 | |
76.16 | |
Ethyl methyl sulfide |
-106°C | |
66°C to 67°C | |
Stench | |
1.4392 | |
MFCD00009268 | |
UN1993 | |
ethyl methyl sulfide, methylthioethane, ethane, methylthio, methyl ethyl sulfide, ethyl methyl sulphide, methyl ethyl sulphide, sulfide, ethyl methyl, unii-mr1gzs62tm, methylthio ethane, methylsulfanyl ethane | |
WXEHBUMAEPOYKP-UHFFFAOYSA-N | |
methylsulfanylethane | |
12230 | |
97% |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H225
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 210-868-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only